Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF434 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552200
Description
Immunogen sequence: GEEAVALVED VQRAPGQQVL DSEKDLKVLM KEMAPLGATR ESLRSQWKQE VQPEEPTFKG SQSSHQRPGE QSEAWLAPQA PRNLPQNTGL H Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 31%.
May be involved in transcriptional regulation.
Specifications
| ZNF434 | |
| Polyclonal | |
| Unconjugated | |
| ZSCAN32 | |
| HCCS5; HCCS-5; Human cervical cancer suppressor gene 5 protein; zinc finger and SCAN domain containing 32; zinc finger and SCAN domain-containing protein 32; Zinc finger protein 434; ZNF434; ZSCAN32 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54925 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NX65 | |
| ZSCAN32 | |
| Recombinant protein corresponding to Human ZNF434. Recombinant protein control fragment (Product #RP-89281). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction