Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF434 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598262
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598262 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598262 Supplier Invitrogen™ Supplier No. PA598262
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF434 Polyclonal Antibody for Western Blot, ELISA

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF434
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZSCAN32
Gene Accession No. Q9NX65
Gene Alias HCCS5; HCCS-5; Human cervical cancer suppressor gene 5 protein; zinc finger and SCAN domain containing 32; zinc finger and SCAN domain-containing protein 32; Zinc finger protein 434; ZNF434; ZSCAN32
Gene Symbols ZSCAN32
Host Species Rabbit
Immunogen Recombinant Human Zinc finger and SCAN domain-containing protein 32 protein (6-248AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54925
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.