Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF435 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541179
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541179 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541179 Supplier Invitrogen™ Supplier No. PA541179
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TKNEELFQKE DMPKDKEFLG EINDRLNKDT PQHPKSKDII ENEGRSEWQQ Sequence homology: Human: 100%.

ZNF435 contains 1 SCAN box domain and 4 C2H2-type zinc fingers. It belongs to the kruppel C2H2-type zinc finger protein family and may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF435
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZSCAN16
Gene Accession No. Q9H4T2
Gene Alias dJ265C24.3; zinc finger and SCAN domain containing 16; zinc finger and SCAN domain-containing protein 16; Zinc finger protein 392; zinc finger protein 435; ZNF392; ZNF435; ZSCAN16
Gene Symbols ZSCAN16
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF435.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80345
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.