Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF435 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552417 Supplier Invitrogen™ Supplier No. PA552417
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLSKHRRTHT GEKPYKCDEC GKAFIQRSHL IGHHRVHTGV KPYKCKECGK DFSGRTGLIQ HQRIHTGEKP YECDECGR Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 73%.

ZNF435 contains 1 SCAN box domain and 4 C2H2-type zinc fingers. It belongs to the kruppel C2H2-type zinc finger protein family and may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF435
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZSCAN16
Gene Accession No. Q9H4T2
Gene Alias dJ265C24.3; zinc finger and SCAN domain containing 16; zinc finger and SCAN domain-containing protein 16; Zinc finger protein 392; zinc finger protein 435; ZNF392; ZNF435; ZSCAN16
Gene Symbols ZSCAN16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF435. Recombinant protein control fragment (Product #RP-89437).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80345
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.