Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF435 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101260
Description
Antibody detects endogenous levels of total ZNF435.
ZNF435 contains 1 SCAN box domain and 4 C2H2-type zinc fingers. It belongs to the kruppel C2H2-type zinc finger protein family and may be involved in transcriptional regulation.
Specifications
ZNF435 | |
Polyclonal | |
Unconjugated | |
ZSCAN16 | |
dJ265C24.3; zinc finger and SCAN domain containing 16; zinc finger and SCAN domain-containing protein 16; Zinc finger protein 392; zinc finger protein 435; ZNF392; ZNF435; ZSCAN16 | |
Rabbit | |
Affinity chromatography | |
RUO | |
80345 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9H4T2 | |
ZSCAN16 | |
A synthesized peptide derived from human ZSCAN16(Accession Q9H4T2), corresponding to amino acid residues E201-L251. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction