Learn More
Invitrogen™ ZNF436 Polyclonal Antibody

Description
Immunogen sequence: SENEVNPKQE ISEDVQFGTT SERPAENAEE NPESEEGFES GDRSERQWGD LTAEEWVSYP LQPVTDLLVH KEVH Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 62%.
Specifications
Specifications
| Antigen | ZNF436 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZNF436 |
| Gene Accession No. | Q9C0F3 |
| Gene Alias | DNA-binding protein; KIAA1710; Zfp46; Zinc finger protein 436; ZNF; ZNF436 |
| Gene Symbols | ZNF436 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.