Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF44 Monoclonal Antibody (4E2)
GREENER_CHOICE

Catalog No. PIMA521976
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521976 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521976 Supplier Invitrogen™ Supplier No. MA521976
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MALCYGTFWG YPKMLEAANL MEGLVDIGPW VTLPRGQPEV LEWGLPKDQD SVAFEDVAVN FTHEEWALLG PSQKNLYRDV MRETIRNLNC IGMKWENQNI DDQ.

ZNF44 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF44
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 4E2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF44
Gene Accession No. P15621
Gene Alias GIOT2; GIOT-2; gonadotropin inducible transcription repressor-2; gonadotropin-inducible ovary transcription repressor 2; gonadotropin-inducible transcription repressor 2; KOX7; Zinc finger protein 44; zinc finger protein 55; Zinc finger protein 58; zinc finger protein KOX7; zinc finger protein ZnFP12; ZNF; ZNF44; ZNF504; ZNF55; ZNF58
Gene Symbols ZNF44
Host Species Mouse
Immunogen ZNF44 (NP_057348.2, 1 a.a. ~ 103 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51710
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.