Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551838
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551838 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551838 Supplier Invitrogen™ Supplier No. PA551838
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HEEWALLGPS QKNLYRDVMR ETIRNLNCIG MKWENQNIDD QHQNLRRNPR CDVVERFGKS KDGSQCGETL SQIRNSIVNK NTPARVDACG SSVNGEVIMG HSSLNCYIRV DTGHKHRECH EYAEK Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 43%.

ZNF44 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF44
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF44
Gene Accession No. P15621
Gene Alias GIOT2; GIOT-2; gonadotropin inducible transcription repressor-2; gonadotropin-inducible ovary transcription repressor 2; gonadotropin-inducible transcription repressor 2; KOX7; Zinc finger protein 44; zinc finger protein 55; Zinc finger protein 58; zinc finger protein KOX7; zinc finger protein ZnFP12; ZNF; ZNF44; ZNF504; ZNF55; ZNF58
Gene Symbols ZNF44
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF44. Recombinant protein control fragment (Product #RP-99754).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51710
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.