Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF44 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544858
Description
Peptide sequence: NLRRNPRCDV VERFGKSKDG SQCGETLSQI RNSIVNKNTP ARVDACGSSV Sequence homology: Human: 100%.
ZNF44 may be involved in transcriptional regulation.
Specifications
| ZNF44 | |
| Polyclonal | |
| Unconjugated | |
| ZNF44 | |
| GIOT2; GIOT-2; gonadotropin inducible transcription repressor-2; gonadotropin-inducible ovary transcription repressor 2; gonadotropin-inducible transcription repressor 2; KOX7; Zinc finger protein 44; zinc finger protein 55; Zinc finger protein 58; zinc finger protein KOX7; zinc finger protein ZnFP12; ZNF; ZNF44; ZNF504; ZNF55; ZNF58 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51710 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P15621 | |
| ZNF44 | |
| Synthetic peptide directed towards the N-terminal of human ZNF44 (aa 106-155). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction