Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF440 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113626 Supplier Invitrogen™ Supplier No. PA5113626
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PMSVSNDGKP SDLPHTFEYV VGHTMERSPM HVRNVGNPSD LPRTFEFMKG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

ZNF440 contains 1 KRAB domain and 12 C2H2-type zinc fingers. ZNF440 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF440
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF440
Gene Accession No. Q8IYI8
Gene Alias Zinc finger protein 440; ZNF440
Gene Symbols ZNF440
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal region of human ZNF440 (aa 542-591).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126070
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.