Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF441 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | ZNF441 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ZNF441 Polyclonal specifically detects ZNF441 in Human samples. It is validated for Western Blot.Specifications
ZNF441 | |
Polyclonal | |
Rabbit | |
Human | |
FLJ38637, zinc finger protein 441 | |
ZNF441 | |
IgG | |
72 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_689568 | |
126068 | |
Synthetic peptide directed towards the N terminal of human ZNF441. Peptide sequence MVERACEIKDNSQCGGPFTQTQDSIVNEKIPGVDPWESSECTDVLMGRSS. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title