Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF442 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570382
Description
This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 85%; Horse: 92%; Human: 100%; Mouse: 86%; Pig: 86%; Rat: 92%.
May be involved in transcriptional regulation.
Specifications
ZNF442 | |
Polyclonal | |
Unconjugated | |
ZNF442 | |
LOC100909622; Zinc finger protein 442; zinc finger protein 442-like; ZNF442 | |
Rabbit | |
Affinity chromatography | |
RUO | |
79973 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H7R0 | |
ZNF442 | |
synthetic peptide directed towards the middle region of human ZNF442. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction