Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF443 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551839 Supplier Invitrogen™ Supplier No. PA551839
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VALEDVAVNF TREEWALLGP CQKNLYKDVM QETIRNLDCV VMKWKDQNIE DQYRYPRKNL RCRMLERFVE SKDGTQCGET SSQIQDSIVT KNTLPGVGPC ESSMRGEKVM GHSSLNCYIR VGA Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 43%.

Zinc finger proteins (ZNFs) bind DNA and, through this binding, regulate gene transcription. Most ZNFs contain conserved C2H2 motifs and are classified as Kruppel-type zinc fingers. For a general description of these proteins, see ZNF91.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF443
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF443
Gene Accession No. Q9Y2A4
Gene Alias Krueppel-type zinc finger protein ZK1; Kruppel-type zinc finger (C2H2); zinc finger protein 443; ZK1; ZNF443
Gene Symbols ZNF443
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF443. Recombinant protein control fragment (Product #RP-99762).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10224
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.