Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF444 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178190AF750
Description
ZNF444 Polyclonal antibody specifically detects ZNF444 in Human samples. It is validated for Western BlotSpecifications
ZNF444 | |
Polyclonal | |
Western Blot | |
HZF3, MGC126498, zinc finger protein 268, zinc finger protein 3, Zinc finger protein HZF3 | |
The immunogen for this product maps to a region between residue 150 and 200 of human Zinc Finger Protein 444 using the numbering given in entry NP_060807.2 (GeneID 55311). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55311 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction