Learn More
ZNF444 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303269AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
ZNF444 has a domain structure and amino acid sequence similar to several zinc finger transcription factors, which strongly suggested that ZNF444 is a transcriptional regulator.ZNF444 has a domain structure and amino acid sequence similar to several zinc finger transcription factors, such as Kruppel-like ZNF191 (MIM 194534).[supplied by OMIM].Specifications
ZNF444 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8N0Y2 | |
ZNF444 | |
Between 200 and 250 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ZNF444 | |
Zinc finger protein 444, EZF2, EZF-2, ZSCAN17 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55311 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.