Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF445 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568542 Supplier Invitrogen™ Supplier No. PA568542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 83%; Dog: 92%; Horse: 92%; Human: 100%; Mouse: 92%; Pig: 85%; Rabbit: 92%; Rat: 92%.

ZNF445 gene ontology annotations related to this gene include DNA-binding transcription factor activity; metal ion binding; nucleic acid binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF445
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF445
Gene Accession No. P59923
Gene Alias AW610627; mszf29; mszf50; RGD1559144; Zfp445; Zinc finger protein 168; zinc finger protein 445; Zinc finger protein with KRAB and SCAN domains 15; ZKSCAN15; ZNF168; Znf445; ZSCAN47
Gene Symbols ZNF445
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ZNF445.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353274
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.