Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF446 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541587 Supplier Invitrogen™ Supplier No. PA541587
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MPSPLGPPCL PVMDPETTLE EPETARLRFR GFCYQEVAGP REALARLREL Sequence homology: Human: 100%; Mouse: 90%; Rat: 90%.

ZNF446 contains a KRAB and three C(2)H(2) zinc fingers. ZNF446 is a transcription repressor when fused to GAL4 DNA-binding domain and co-transfected with VP-16. Overexpression of ZNF446 in COS-7 cells inhibits the transcriptional activities of SRE.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF446
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF446
Gene Accession No. Q9NWS9
Gene Alias FLJ20626; Zinc finger protein 446; zinc finger protein with KRAB and SCAN domains 20; ZKSCAN20; ZNF446; ZSCAN30; ZSCAN52
Gene Symbols ZNF446
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF446 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55663
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.