Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF447 Antibody (OTI2H7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274951B
Description
ZNF447 Monoclonal specifically detects ZNF447 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ZNF447 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
ZSCAN18 | |
Full length recombinant protein of human ZNF447 (NP_076415) produced in HEK293T cells. | |
0.1 mL | |
65982.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2H7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DKFZp586B1122, FLJ12895, FLJ38919, FLJ44152, MGC4074, MGC8682, zinc finger and SCAN domain containing 18, zinc finger and SCAN domain-containing protein 18, Zinc finger protein 447MGC2427, ZNF447 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction