Learn More
Invitrogen™ ZNF447 Monoclonal Antibody (2H1)

Description
Peptide Sequence: MLPLEKAFAS PRSSPAPPDL PTPGSAAGVQ QEEPETIPER TPADLEFSRL RFREFVYQEA AGPHQTLARL HELCRQWLMP EARSKEQMLE LLVLEQFLGI LPDKVRPWVV AQYPESCKKA ASLVEGLADV LEEPGMLLGS PAGSSSILSD GVYERHMDPL LLPGELASPS QALGAGEIPA PSETPWLSPD PLFLEQRRVR EAKTEEDGPA NTEQKLKSFP EDPQHLGEWG HLDPAEENLK SYRKLLLWGY QLSQPDAASR LDTEELRLVE RDPQGSSLPE GGRRQESAGC ACEEAAPAGV LPELPTEAPP GDALADPPSG TTEEEEEQPG KAPDPQDPQD AESDSATGSQ RQSVIQQPAP DRGTAKLGTK RPHPEDGDEQ SLEGVSSSGD SAGLEAGQGP GADEPGLSRG KPYACGECGE AFAWLSHLME HHSSHGGRKR YACQGCWKTF HFSLALAEHQ KTHEKEKSYA LGGARGPQPS TREAQAGARA GGPPESVEGE APPAPPEAQR.
Specifications
Specifications
| Antigen | ZNF447 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2H1 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZSCAN18 |
| Gene Accession No. | Q8TBC5 |
| Gene Alias | EG232875; RGD1560671; similar to hypothetical protein FLJ12895; zinc finger and SCAN domain containing 18; zinc finger and SCAN domain-containing protein 18; Zinc finger protein 447; ZNF447; ZSCAN18 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.