Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF449 Antibody (OTI4C6), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274952FR
Description
ZNF449 Monoclonal specifically detects ZNF449 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ZNF449 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
ZNF449 | |
Full length recombinant protein of human ZNF449 (NP_689908) produced in E.coli. | |
0.1 mL | |
203523.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4C6 | |
Western Blot, Immunohistochemistry | |
HZF1Zinc finger protein HZF1, KIAA2007, zinc finger protein 1, zinc finger protein 266 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction