Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567374
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567374 Supplier Invitrogen™ Supplier No. PA567374
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HQRVHADDEGD KDFPSSEDSH RKTR Highest antigen sequence identity to the following orthologs - mouse 60%, rat 64%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF45
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF45
Gene Accession No. Q02386
Gene Alias BRC1744; KOX5; Zinc finger protein 13; zinc finger protein 45; zinc finger protein 45 (a Kruppel-associated box (KRAB) domain polypeptide); zinc finger protein KOX5; ZNF13; ZNF45
Gene Symbols ZNF45
Host Species Rabbit
Immunogen Recombinant Human ZNF45. Recombinant protein control fragment (Product #RP-108035).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7596
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.