Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF45 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA558466
Description
Immunogen sequence: LWMMKMATQR DNSSGAKNLK EMETLQEVGL RYLPHEELFC SQIWQQITRE LIKYQDSVVN IQRTGCQLEK RDDLHYKDEG FSNQSSHLQV HR Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 50%.
May be involved in transcriptional regulation.
Specifications
ZNF45 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q02386 | |
ZNF45 | |
Recombit protein corresponding to Human ZNF45 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
ZNF45 | |
KOX5, ZNF13, BRC1744, zinc finger protein 13, zinc finger protein 45 (a Kruppel-associated box (KRAB) domain polypeptide), zinc finger protein KOX5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7596 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction