Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF451 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116621
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116621 Supplier Invitrogen™ Supplier No. PA5116621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cellular location: Nucleus Immunogen sequence: SSPRGKWQCR ICEDMFDSQE YVKQHCMSLA SHKFHRYSCA HCRKPFHKIE TLYRHCQDEH DNEIKIKYFC GLCDLIFNVE EAFLSHYEEH HSIDYVFVSE KTETSIKTED DFPVIETSNQ LTCGCRESYI CKVNRKEDYS RCLQIMLDKG KLWFRCSLCS ATAQNLTDMN THIHQVHKEK SDEEEQQYVI KCGTCTKAFH DPESAQQHFH RKHCFLQKPS VAHFGSEKSN LYKFTASASH TERKLKQAIN YSKSLDMEKG VENDLSYQNI EEEIVELPDL DYLRTMTHIV FVDFDNWSNF F.

E3 SUMO-protein ligase; has a preference for SUMO2 and SUMO3 and facilitates UBE2I/UBC9-mediated sumoylation of target proteins (PubMed:26524493, PubMed:26524494). Plays a role in protein SUMO2 modification in response to stress caused by DNA damage and by proteasome inhibitors (in vitro). Required for MCM4 sumoylation. Has no activity with SUMO1 (PubMed:26524493). Preferentially transfers an additional SUMO2 chain onto the SUMO2 consensus site 'Lys-11' (PubMed:26524493). Negatively regulates transcriptional activation mediated by the SMAD4 complex in response to TGF-beta signaling. Inhibits EP300-mediated acetylation of histone H3 at 'Lys-9' (PubMed:24324267). Plays a role in regulating the transcription of AR targets (PubMed:18656483). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF451
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZNF451
Gene Accession No. Q8C0P7, Q9Y4E5
Gene Alias 4930515K21Rik; 4933435G09Rik; AI596398; Coactivator for steroid receptors; COASTER; dJ417I1.1; E3 SUMO-protein ligase ZNF451; E3 SUMO-protein ligase ZNF451; zinc finger protein 451; E3 SUMO-protein transferase ZNF451; KIAA0576; Kiaa0576-hp; KIAA1702; mKIAA1702; Zfp451; Zinc finger protein 451; Zinc finger protein 451-like protein; Znf451
Gene Symbols Zfp451, ZNF451
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26036, 316312, 98403
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.