Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF451 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572615
Description
E3 SUMO-protein ligase; has a preference for SUMO2 and SUMO3 and facilitates UBE2I/UBC9-mediated sumoylation of target proteins (PubMed:26524493, PubMed:26524494). Plays a role in protein SUMO2 modification in response to stress caused by DNA damage and by proteasome inhibitors (in vitro). Required for MCM4 sumoylation (By similarity). Has no activity with SUMO1 (PubMed:26524493). Preferentially transfers an additional SUMO2 chain onto the SUMO2 consensus site ′Lys-11′ (PubMed:26524493). Negatively regulates transcriptional activation mediated by the SMAD4 complex in response to TGF-beta signaling. Inhibits EP300-mediated acetylation of histone H3 at ′Lys-9′ (PubMed:24324267). Plays a role in regulating the transcription of AR targets (PubMed:18656483).
Specifications
ZNF451 | |
Polyclonal | |
Unconjugated | |
ZNF451 | |
Zinc finger protein 451; Coactivator for steroid receptors; ZNF451; COASTER; KIAA0576; KIAA1702 | |
Rabbit | |
Protein A | |
RUO | |
26036 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.4 mg/mL | |
PBS with 0.09% sodium azide | |
Q9Y4E5 | |
ZNF451 | |
KLH conjugated synthetic peptide between 81-110 amino acids from the N-terminal region of human ZNF451 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction