Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF460 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562715
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562715 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562715 Supplier Invitrogen™ Supplier No. PA562715
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKMSLEHEGL ATADGICSMM IQNQVSPEDA LYGFDSYGPV TDSLIHEGEN SYKFEEMFNE NCFLVQHEQI LPRVKPYDC Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 29%.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppeltype DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF460 (zinc finger protein 460), also known as HZF8 or ZNF272, is a 562 amino acid nuclear protein that is ubiquitously expressed at low levels and expressed at high levels in pancreas and liver. Belonging to the Krueppel C2 H2-type zinc-finger protein family, ZNF460 contains 11 C2 H2-type zinc fingers and a KRAB domain. Like other zinc-fingers, ZNF460 is also suggested to be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF460
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF460
Gene Accession No. Q14592
Gene Alias HZF8; Zinc finger protein 272; zinc finger protein 460; Zinc finger protein HZF8; ZNF272; ZNF460
Gene Symbols ZNF460
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF460. Recombinant protein control fragment (Product #RP-91700).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10794
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.