Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF460 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110602
Description
Immunogen sequence: SEMQEYFLRP GTDPQSEKLH GKMSLEHEGL ATADGICSMM IQNQVSPEDA LYGFDSYGPV TDSLIHEGEN SYKFEEMFNE NCFLVQHEQI LPRVKPYDCP ECGKAFGKSK H.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppeltype DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF460 (zinc finger protein 460), also known as HZF8 or ZNF272, is a 562 amino acid nuclear protein that is ubiquitously expressed at low levels and expressed at high levels in pancreas and liver. Belonging to the Krueppel C2 H2-type zinc-finger protein family, ZNF460 contains 11 C2 H2-type zinc fingers and a KRAB domain. Like other zinc-fingers, ZNF460 is also suggested to be involved in transcriptional regulation.
Specifications
ZNF460 | |
Polyclonal | |
Unconjugated | |
ZNF460 | |
HZF8; Zinc finger protein 272; zinc finger protein 460; Zinc finger protein HZF8; ZNF272; ZNF460 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10794 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.22 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q14592 | |
ZNF460 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 100-210 of human ZNF460 (NP_006626.3). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction