Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF460 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734845 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4845 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4845 Supplier Proteintech Group Inc Supplier No. 252991AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppeltype DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF460 (zinc finger protein 460), also known as HZF8 or ZNF272, is a 562 amino acid nuclear protein that is ubiquitously expressed at low levels and expressed at high levels in pancreas and liver. Belonging to the Krueppel C2 H2-type zinc-finger protein family, ZNF460 contains 11 C2 H2-type zinc fingers and a KRAB domain. Like other zinc-fingers, ZNF460 is also suggested to be involved in transcriptional regulation.

Specifications

Antigen ZNF460
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ZNF460
Gene Accession No. Q14592
Gene Alias HZF8, Zinc finger protein 272, zinc finger protein 460, Zinc finger protein HZF8, ZNF272, ZNF460
Gene Symbols ZNF460
Host Species Rabbit
Immunogen ZNF460 Fusion Protein Ag18106
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10794
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.