Learn More
Invitrogen™ ZNF461 Monoclonal Antibody (4G11)

Description
Peptide Sequence: ELSQWVNMEE FKSHSPERSI FSAIWEGNCH FEQHQGQEEG YFRQLMINHE NMPIFSQHTL LTQEFYDREK ISECKKCRKI FSYHLFFSHH KRTHSKEL.
Specifications
Specifications
| Antigen | ZNF461 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4G11 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZNF461 |
| Gene Accession No. | Q8TAF7 |
| Gene Alias | GIOT1; GIOT-1; Gonadotropin-inducible ovary transcription repressor 1; HZF28; zinc finger protein 28; zinc finger protein 461; ZNF461 |
| Gene Symbols | ZNF461 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.