Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF461 Polyclonal Antibody
ZNF461 Polyclonal Antibody
Supplier: Thermo Scientific PA569053
Description
ZNF461 Polyclonal Antibody for Western Blot

Specifications
ZNF461 | |
Polyclonal | |
Unconjugated | |
ZNF461 | |
GIOT1; GIOT-1; Gonadotropin-inducible ovary transcription repressor 1; HZF28; zinc finger protein 28; zinc finger protein 461; ZNF461 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
92283 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TAF7 | |
ZNF461 | |
synthetic peptide directed towards the C-terminal region of human ZNF461. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction