Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF467 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568446
Description
ZNF467 Polyclonal Antibody for IHC (P)
Transcription factor that promotes adipocyte differentiation and suppresses osteoblast differentiation in the bone marrow. Enhances the osteoclast-supporting ability of stromal cells. Binds with STAT3 the consensus sequence 5'-CTTCTGGGAAGA-3' of the acute phase response element (APRE). Transactivates several promoters including FOS, OSM and PPARG. Recruits a histone deacetylase complex.
Specifications
ZNF467 | |
Polyclonal | |
Unconjugated | |
ZNF467 | |
EZI; Zfp467; Zinc finger protein 467; zinc finger protein EZI; ZNF467 | |
Rabbit | |
affinity chromatography | |
RUO | |
168544 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q7Z7K2 | |
ZNF467 | |
Synthesized peptide derived from human ZNF467. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction