Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF471 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564281
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564281 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564281 Supplier Invitrogen™ Supplier No. PA564281
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESIYVTQELP LKQFMYDDAC MEGITSYGLE CS Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 44%.

ZNF471 (Zinc Finger Protein 471) is a Protein Coding gene. Among its related pathways are Gene Expression. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and DNA-binding transcription factor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF471
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF471
Gene Accession No. Q9BX82
Gene Alias ERP1; EZFIT-related protein 1; KIAA1396; Z1971; zinc finger protein 471; ZNF471
Gene Symbols ZNF471
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF471. Recombinant protein control fragment (Product #RP-103636).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57573
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.