Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF473 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552069
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552069 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552069 Supplier Invitrogen™ Supplier No. PA552069
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSPEATSPDV TETKNSPLME DFFEEGFSQE IIEMLSKDGF WNSNFGEACI EDTWLDSLLG DPESLLRSDI ATNGESPTEC KSHELKRGLS PVSTVSTGED SMVHNVSEKT LTPAKSKEYR GEFFSYSDHS Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 45%.

Involved in histone 3'-end pre-mRNA processing by associating with U7 snRNP and interacting with SLBP/pre-mRNA complex. Increases histone 3'-end pre-mRNA processing but has no effect on U7 snRNP levels, when overexpressed. Required for cell cycle progression from G1 to S phases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF473
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF473
Gene Accession No. Q8WTR7
Gene Alias HZFP100; KIAA1141; ZFP100; Zfp-100; Zinc finger protein 100 homolog; zinc finger protein 473; ZN473; ZNF473
Gene Symbols ZNF473
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF473. Recombinant protein control fragment (Product #RP-99747).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25888
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.