Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF48 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554891
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554891 Supplier Invitrogen™ Supplier No. PA554891
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLGSENVISQ PNEFEHTPQE DDLGFKEEDL APDHEVGNVS LKPEGIQNWD DLWVQREGLG KPQPRDRGPR LLGEPRWGQA SSDRA Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 68%.

ZNF48, also named as zinc finger protein 48, is 618 amino acid protein, which contains twelves C2H2-type zinc fingers and belongs to the Krueppel C2H2-type zinc-finger protein family. ZNF48 localizes in the nucleus and may be involves in transcription regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF48
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF48
Gene Accession No. Q96MX3
Gene Alias Zinc finger protein 48; zinc finger protein 553; ZNF48; ZNF553
Gene Symbols ZNF48
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF48. Recombinant protein control fragment (Product #RP-93626).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197407
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.