Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF48 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734919 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4919 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4919 Supplier Proteintech Group Inc Supplier No. 254631AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF48, also named as zinc finger protein 48, is 618 amino acid protein, which contains twelves C2H2-type zinc fingers and belongs to the Krueppel C2H2-type zinc-finger protein family. ZNF48 localizes in the nucleus and may be involves in transcription regulation.

Specifications

Antigen ZNF48
Applications Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ZNF48
Gene Accession No. Q96MX3
Gene Alias zinc finger protein 48, Zinc finger protein 553, ZNF48, ZNF553
Gene Symbols ZNF48
Host Species Rabbit
Immunogen ZNF48 Fusion Protein Ag22208
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197407
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.