Learn More
ZNF480 Mouse anti-Human, Clone: OTI8E3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF812668

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The function of this protein remains unknown.Specifications
ZNF480 | |
Monoclonal | |
Unconjugated | |
ZNF480 | |
zinc finger protein 480 | |
Mouse | |
Affinity Chromatography | |
RUO | |
147657 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8E3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8WV37 | |
ZNF480 | |
Full length human recombit protein of human ZNF480 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.