Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF485 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $499.50
Specifications
Antigen | ZNF485 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18040020
![]() |
Novus Biologicals
NBP18040020UL |
20 μL |
Each for $158.00
|
|
|||||
NBP180400
![]() |
Novus Biologicals
NBP180400 |
100 μL |
Each for $499.50
|
|
|||||
Description
ZNF485 Polyclonal specifically detects ZNF485 in Human samples. It is validated for Western Blot.Specifications
ZNF485 | |
Polyclonal | |
Rabbit | |
NP_660355 | |
220992 | |
Synthetic peptide directed towards the C terminal of human ZNF485. Peptide sequence: RHSSGLVEHQRLHTGEKPYKCNECGKAFPRSSALKQHKKIHNKERAMKCS | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
zinc finger protein 227 | |
ZNF485 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title