Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF485 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563708
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563708 Supplier Invitrogen™ Supplier No. PA563708
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEPWTEVREA PSGTHAVEDY WFETKMSALK QSTSEASVLG ERTKSVMMEK GLDWEGRSST EK Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 29%.

ZNF485 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF485
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF485
Gene Accession No. Q8NCK3
Gene Alias Zinc finger protein 485; Zinc finger protein 93 (Zinc finger protein HTF34); ZNF485
Gene Symbols ZNF485
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF485. Recombinant protein control fragment (Product #RP-101960).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220992
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.