Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF486 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ZNF486 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ZNF486 Polyclonal specifically detects ZNF486 in Human samples. It is validated for Western Blot.Specifications
ZNF486 | |
Polyclonal | |
Rabbit | |
NP_443084 | |
90649 | |
Synthetic peptide directed towards the C terminal of human ZNF486The immunogen for this antibody is ZNF486. Peptide sequence HRRTHTGEKPYKCEECGKAYTTSSNLTEHKTTHTGEKPYKCKECGKAFNW. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
KOX16MGC57283, Zfp612, zinc finger protein 23, zinc finger protein 23 (KOX 16), zinc finger protein 32, Zinc finger protein 359ZNF612kruppel-like zinc finger factor X31, Zinc finger protein 612, Zinc finger protein KOX16, ZNF359 | |
ZNF486 | |
IgG | |
54 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title