Learn More
Invitrogen™ ZNF486 Polyclonal Antibody

Description
This antibody may cross-react with ZNF506 protein. Recommended positive controls: SK-N-SH and IMR-32 cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ZNF486 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% Thimerosal; pH 7 |
| Gene | ZNF486 |
| Gene Accession No. | Q96H40 |
| Gene Alias | KRAB box only protein 2; KRAB domain only 2; KRAB domain only protein 2; KRBO2; Zinc finger protein 486; ZNF486 |
| Gene Symbols | ZNF486 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.