Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF486 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ZNF486 |
---|---|
Concentration | 0.26 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2982
![]() ![]() |
Proteintech Group Inc
205991AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
ZNF486 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 2 C2H2-type zinc fingers and 1 KRAB domain. ZNF486 may be involved in transcriptional regulation.Specifications
ZNF486 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q96H40 | |
90649 | |
ZNF486 Fusion Protein Ag14567 | |
Primary | |
-20°C | |
ZNF486 |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HCG1999761, isoform CRA_b, KRBO2, zinc finger protein 486, ZNF486 | |
ZNF486 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title