Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF488 Antibody, Novus Biologicals™
SDP

Catalog No. p-7109840 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP180174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP180174 Supplier Novus Biologicals Supplier No. NBP180174
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF488 Polyclonal specifically detects ZNF488 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZNF488
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_694579
Gene Alias FDZF2Zinc finger protein FDZF2, zinc finger protein 230
Gene Symbols ZNF488
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal of human ZNF488. Peptide sequence SDLVFHMRSHHKKEHAGPDPHSQKRREEALACPVCQEHFRERHHLSRHMT.
Molecular Weight of Antigen 37 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118738
Test Specificity Rat: 79%; Mouse: 79%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.