Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF488 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111068
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111068 Supplier Invitrogen™ Supplier No. PA5111068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSGLLNTTDL ACWGRLSTPK LLVGDLWNLQ ALPQNAPLCS TFLGAPTLWL EHTQAQVPPP SSSSTTSWAL LPPTLTSLGL STQNWCAKC Highest antigen sequence indentity to the following orthologs: Mouse - 64%, Rat - 62%.

Transcriptional repressor. Plays a role in oligodendrocyte differentiation, together with OLIG2. Mediates Notch signaling-activated formation of oligodendrocyte precursors. Promotes differentiation of adult neural stem progenitor cells (NSPCs) into mature oligodendrocytes and contributes to remyelination following nerve injury. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF488
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF488
Gene Accession No. Q96MN9
Gene Alias Gm1206; Zfp488; Zinc finger protein 488; Znf488
Gene Symbols ZNF488
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ZNF488. Recombinant protein control fragment (Product #RP-105913).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118738
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.