Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF491 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ZNF491 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ZNF491 Polyclonal specifically detects ZNF491 in Human samples. It is validated for Western Blot.Specifications
ZNF491 | |
Polyclonal | |
Rabbit | |
Human | |
FLJ34791, MGC126639, zinc finger protein 491 | |
ZNF491 | |
IgG | |
51 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_689569 | |
126069 | |
Synthetic peptide directed towards the N terminal of human ZNF491. Peptide Sequence: SFNRNIRTDTGHQPHKCQKFLEKPYKHKQRRKALSHSHCFRTHERPHTRE | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title