Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF496 Polyclonal Antibody
ZNF496 Polyclonal Antibody
Supplier: Thermo Scientific PA5103348
Description
ZNF496 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
ZNF496 | |
Polyclonal | |
Unconjugated | |
ZNF496 | |
MGC15548; NIZP1; Nizp11; NSD1 (nuclear receptor binding SET-domain containing 1)-interacting zinc finger protein 1; NSD1-interacting zinc finger protein 1; Zfp496; zinc finger protein 496; Zinc finger protein with KRAB and SCAN domains 17; zinc finger with KRAB and SCAN domains 17; Zkscan17; ZNF496; ZSCAN49 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
268417, 84838 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5SXI5, Q96IT1 | |
Zkscan17, ZNF496 | |
A synthesized peptide derived from human ZNF496(Accession Q96IT1), corresponding to amino acid residues L5-G55. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction