Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF496 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540311
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540311 Supplier Invitrogen™ Supplier No. PA540311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QEKPHECSVC GELFSDSEDL DGHLESHEAQ KPYRCGACGK SFRLNSHLLS Sequence homology: Cow: 86%; Dog: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 93%; Rat: 93%; Sheep: 86%.

ZNF496 contains a SCAN box, a KRAB-A domain, and four consensus C2H2-type zinc fingers preceded by a unique finger derivative, referred to herein as the C2HR motif. The C2HR motif functions to mediate protein-protein interaction with the cysteine-rich (C5HCH) domain of NSD1 in a Zn(II)-dependent fashion, and when tethered to RNA polymerase II promoters, represses transcription in an NSD1-dependent manner. ZNF496 contains a novel type of zinc finger motif that functions as a docking site for NSD1 and is more than just a degenerate evolutionary remnant of a C2H2 motif.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF496
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF496
Gene Accession No. Q96IT1
Gene Alias MGC15548; NIZP1; Nizp11; NSD1 (nuclear receptor binding SET-domain containing 1)-interacting zinc finger protein 1; NSD1-interacting zinc finger protein 1; Zfp496; zinc finger protein 496; Zinc finger protein with KRAB and SCAN domains 17; zinc finger with KRAB and SCAN domains 17; Zkscan17; ZNF496; ZSCAN49
Gene Symbols ZNF496
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZNF496.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84838
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.