Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF496 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144735
Description
Immunogen sequence: SYVCPNCGKI FRWRVNFIRH LRSRREQEKP HECSVCGELF SDSEDLDGHL ESHEAQKPYR CGACGKSFRL NSHLLSHRRI HLQPDRLQPV EKREQAASED ADKGPKEPLE NGKAKLSFQC CECGKAFQRH DHLARHRSHF HLKD Antigen sequence identity: Mouse 78%; Rat 74%.
ZNF496 contains a SCAN box, a KRAB-A domain, and four consensus C2H2-type zinc fingers preceded by a unique finger derivative, referred to herein as the C2HR motif. The C2HR motif functions to mediate protein-protein interaction with the cysteine-rich (C5HCH) domain of NSD1 in a Zn(II)-dependent fashion, and when tethered to RNA polymerase II promoters, represses transcription in an NSD1-dependent manner. ZNF496 contains a novel type of zinc finger motif that functions as a docking site for NSD1 and is more than just a degenerate evolutionary remnant of a C2H2 motif.
Specifications
ZNF496 | |
Polyclonal | |
Unconjugated | |
ZNF496 | |
MGC15548; NIZP1; Nizp11; NSD1 (nuclear receptor binding SET-domain containing 1)-interacting zinc finger protein 1; NSD1-interacting zinc finger protein 1; Zfp496; zinc finger protein 496; Zinc finger protein with KRAB and SCAN domains 17; zinc finger with KRAB and SCAN domains 17; Zkscan17; ZNF496; ZSCAN49 | |
Rabbit | |
Affinity chromatography | |
RUO | |
84838 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96IT1 | |
ZNF496 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 405-548. Recombinant protein control fragment (Product #RP-109712). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction