Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF498 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599236
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599236 Supplier Invitrogen™ Supplier No. PA599236
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZNF498.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. As a member of the Krüppel C2H2-type zinc-finger protein family, ZNF498 (zinc finger protein 498), also known as Zinc finger and SCAN domain-containing protein 25 (ZSCAN25), is a 544 amino acid protein. Localized to the nucleus, ZNF498 contains seven C2H2-type zinc fingers and one SCAN box domain. Due to the presence of these domains, ZNF498 is believed to be involved in transcriptional regulation. ZNF498 exists as 4 isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF498
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZSCAN25
Gene Accession No. Q6NSZ9
Gene Alias EG666311; Zfp498; zinc finger and SCAN domain containing 25; zinc finger and SCAN domain-containing protein 25; Zinc finger protein 498; Znf498; Zscan25
Gene Symbols ZSCAN25
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZSCAN25(Accession Q6NSZ9), corresponding to amino acid residues S342-K392.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221785, 666311
Target Species Human, Mouse, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.