Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537087
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537087 Supplier Thermo Scientific Supplier No. PA537087
Add to Cart
Add to Cart

ZNF50 Polyclonal Antibody

ZNF50 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF50
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ZSCAN22
Gene Accession No. P10073
Gene Alias AW227621; D530006B18Rik; GLI-Kruppel family member HKR2; HKR2; Krueppel-related zinc finger protein 2; LOC102553002; Protein HKR2; zinc finger and SCAN domain containing 22; zinc finger and SCAN domain-containing protein 22; Zinc finger protein 50; zinc finger protein 8-like; ZNF50; ZSCAN22
Gene Symbols ZSCAN22
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 363-408 of human ZNF50.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102553002, 232878, 342945
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.