Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF500 Antibody (OTI2D2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246454
Description
Ensure accurate, reproducible results in Western Blot
ZNF500 Monoclonal specifically detects ZNF500 in Human samples. It is validated for Western Blot.Specifications
ZNF500 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
C2H2-171, RP58Zinc finger and BTB domain-containing protein 18, TAZ1, TAZ-1Zinc finger protein C2H2-171, Transcriptional repressor RP58, Translin-associated zinc finger protein 1, ZBTB1858 kDa repressor protein, zinc finger protein 238 | |
Mouse | |
53.5 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
OTI2D2 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
ZNF500 | |
Human recombinant protein fragment corresponding to amino acids 102-325 of human ZNF500 (NP_067678) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
26048 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction