Learn More
ZNF500 Mouse anti-Human, Clone: OTI4G10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF803375

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ZNF500 is a new candidate transcription factor. It is derived from an annotated genomic sequence (NT_037887) using a gene prediction method.Specifications
ZNF500 | |
Monoclonal | |
Unconjugated | |
ZNF500 | |
ZKSCAN18, ZSCAN50 | |
Mouse | |
Affinity Chromatography | |
RUO | |
26048 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4G10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O60304 | |
ZNF500 | |
Human recombit protein fragment corresponding to amino acids 102-325 of human ZNF500 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.